Assay Details
| Gene Symbol: akt2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212815.2 | NP_997980.1 | 9 - 10 | 1012 | 111 | |
| GenBank mRNA | BC046892.1 | - | 9 - 10 | 1013 | 111 | |
| BC068106.1 | - | 9 - 10 | 980 | 111 | ||
| BC165215.1 | - | 8 - 9 | 862 | 111 | ||
More Information
Set Membership: |
|

akt2l
Genomic Map