Assay Details
| Gene Symbol: gatad2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212916.1 | NP_998081.1 | 10 - 11 | 1892 | 131 | |
| XM_005170444.4 | XP_005170501.1 | 10 - 11 | 2204 | 131 | ||
| XM_005170445.4 | XP_005170502.1 | 10 - 11 | 2183 | 131 | ||
| XM_005170447.4 | XP_005170504.2 | 10 - 11 | 1934 | 131 | ||
| XM_005170448.4 | XP_005170505.1 | 10 - 11 | 1958 | 131 | ||
| GenBank mRNA | BC067707.1 | - | 10 - 11 | 1892 | 131 | |
More Information
Set Membership: |
|

gatad2ab
Genomic Map