Assay Details
| Gene Symbol: gatad2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212916.1 | NP_998081.1 | 9 - 10 | 1672 | 101 | |
| XM_005170444.4 | XP_005170501.1 | 9 - 10 | 1984 | 101 | ||
| XM_005170445.4 | XP_005170502.1 | 9 - 10 | 1963 | 101 | ||
| XM_005170446.4 | XP_005170503.1 | 9 - 10 | 1985 | 101 | ||
| XM_005170447.4 | XP_005170504.2 | 9 - 10 | 1714 | 101 | ||
| XM_005170448.4 | XP_005170505.1 | 9 - 10 | 1738 | 101 | ||
| GenBank mRNA | BC067707.1 | - | 9 - 10 | 1672 | 101 | |
More Information
Set Membership: |
|

gatad2ab
Genomic Map