Assay Details
| Gene Symbol: lgals3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214693.2 | NP_999858.2 | 5 - 6 | 580 | 108 | |
| GenBank mRNA | AY421705.1 | - | 5 - 6 | 569 | 108 | |
| BC107990.1 | - | 5 - 6 | 580 | 108 | ||
More Information
Set Membership: |
|

lgals3b
Genomic Map