Assay Details
| Gene Symbol: rab6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328529.1 | NP_001315458.1 | 1 - 2 | 270 | 98 | |
| NM_001328530.1 | NP_001315459.1 | 1 - 2 | 270 | 98 | ||
| XM_009305566.1 | XP_009303841.1 | 1 - 2 | 270 | 98 | ||
| XM_009305567.2 | XP_009303842.1 | 1 - 2 | 270 | 98 | ||
| GenBank mRNA | BC044491.1 | - | 1 - 2 | 251 | 98 | |
| BC157379.1 | - | 1 - 2 | 116 | 98 | ||
| EH453941.1 | - | 1 - 2 | 228 | 98 | ||
More Information
Set Membership: |
|

rab6a
Genomic Map