Assay Details
| Gene Symbol: ralbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002123.1 | NP_001002123.1 | 490 | 65 | ||
| GenBank mRNA | BC071460.1 | - | 490 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ralbb| Gene Symbol: ralbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002123.1 | NP_001002123.1 | 490 | 65 | ||
| GenBank mRNA | BC071460.1 | - | 490 | 65 | ||
Set Membership: |
|