Assay Details
| Gene Symbol: tha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002176.1 | NP_001002176.1 | 6 - 7 | 1017 | 102 | |
| XM_009306814.3 | XP_009305089.1 | 5 - 6 | 956 | 102 | ||
| XM_021480050.1 | XP_021335725.1 | 6 - 7 | 762 | 102 | ||
| GenBank mRNA | BC072718.1 | - | 6 - 7 | 1017 | 102 | |
| BC154201.1 | - | 6 - 7 | 1003 | 102 | ||
More Information
Set Membership: |
|

tha1
Genomic Map