Assay Details
| Gene Symbol: litaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002184.1 | NP_001002184.1 | 3 - 4 | 414 | 95 | |
| XM_009299704.3 | XP_009297979.1 | 3 - 4 | 677 | 95 | ||
| GenBank mRNA | BC074085.1 | - | 3 - 4 | 414 | 95 | |
More Information
Set Membership: |
|

litaf
Genomic Map