Assay Details
| Gene Symbol: plpp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002211.1 | NP_001002211.1 | 5 - 6 | 593 | 71 | |
| XM_005155296.4 | XP_005155353.1 | 5 - 6 | 654 | 71 | ||
| XM_005155298.3 | XP_005155355.1 | 5 - 6 | 590 | 71 | ||
| GenBank mRNA | BC074073.1 | - | 5 - 6 | 593 | 71 | |
| BC164605.1 | - | 4 - 5 | 375 | 71 | ||
More Information
Set Membership: |
|

plpp5
Genomic Map