Assay Details
| Gene Symbol: get3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002298.2 | NP_001002298.1 | 3 - 4 | 577 | 97 | |
| GenBank mRNA | BC071461.1 | - | 3 - 4 | 577 | 97 | |
| EH509131.1 | - | 3 - 4 | 554 | 97 | ||
More Information
Set Membership: |
|

get3
Genomic Map