Assay Details
| Gene Symbol: get3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002298.2 | NP_001002298.1 | 6 - 7 | 1042 | 80 | |
| GenBank mRNA | BC071461.1 | - | 6 - 7 | 1042 | 80 | |
More Information
Set Membership: |
|

get3
Genomic Map