Assay Details
| Gene Symbol: hax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002337.1 | NP_001002337.1 | 2 - 3 | 368 | 63 | |
| GenBank mRNA | BC075929.1 | - | 2 - 3 | 368 | 63 | |
More Information
Set Membership: |
|

hax1
Genomic Map