Assay Details
| Gene Symbol: calca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002471.1 | NP_001002471.1 | 2 - 3 | 189 | 67 | |
| XM_005172267.4 | XP_005172324.1 | 2 - 3 | 242 | 67 | ||
| GenBank mRNA | BC076343.1 | - | 2 - 3 | 189 | 67 | |
| BC164929.1 | - | 1 - 2 | 124 | 67 | ||
| DQ406589.1 | - | 1 - 2 | 90 | 67 | ||
| DQ406590.1 | - | 1 - 2 | 90 | 67 | ||
| JN837104.1 | - | 1 - 2 | 69 | 67 | ||
More Information
Set Membership: |
|

calca
Genomic Map