Assay Details
| Gene Symbol: clic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002561.1 | NP_001002561.1 | 1 - 2 | 199 | 63 | |
| GenBank mRNA | BC076239.1 | - | 1 - 2 | 199 | 63 | |
More Information
Set Membership: |
|

clic2
Genomic Map