Assay Details
| Gene Symbol: sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002605.1 | NP_001002605.1 | 8 - 9 | 870 | 63 | |
| XM_005170143.4 | XP_005170200.1 | 8 - 9 | 779 | 63 | ||
| GenBank mRNA | BC075987.1 | - | 8 - 9 | 870 | 63 | |
More Information
Set Membership: |
|

sirt5
Genomic Map