Assay Details
| Gene Symbol: dpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002683.1 | NP_001002683.1 | 13 - 14 | 1554 | 64 | |
| GenBank mRNA | BC075771.1 | - | 13 - 14 | 1554 | 64 | |
More Information
Set Membership: |
|

dpp3
Genomic Map