Assay Details
| Gene Symbol: tyrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002749.2 | NP_001002749.1 | 3 - 4 | 790 | 93 | |
| GenBank mRNA | BC076406.1 | - | 3 - 4 | 758 | 93 | |
More Information
Set Membership: |
|

tyrp1b
Genomic Map