Assay Details
| Gene Symbol: tyrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002749.2 | NP_001002749.1 | 6 - 7 | 1346 | 84 | |
| GenBank mRNA | BC076406.1 | - | 6 - 7 | 1314 | 84 | |
More Information
Set Membership: |
|

tyrp1b
Genomic Map