Assay Details
| Gene Symbol: gnl3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002875.1 | NP_001002875.1 | 8 - 9 | 842 | 106 | |
| GenBank mRNA | AY648737.1 | - | 8 - 9 | 842 | 106 | |
| BC078411.1 | - | 8 - 9 | 765 | 106 | ||
| BC091975.1 | - | 8 - 9 | 789 | 106 | ||
| BC164275.1 | - | 7 - 8 | 781 | 106 | ||
More Information
Set Membership: |
|

gnl3l
Genomic Map