Assay Details
| Gene Symbol: nadk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003465.2 | NP_001003465.2 | 7 - 8 | 1103 | 64 | |
| XM_005161165.4 | XP_005161222.1 | 7 - 8 | 1129 | 64 | ||
| XM_005161166.4 | XP_005161223.1 | 6 - 7 | 1402 | 64 | ||
| XM_009295302.3 | XP_009293577.1 | 7 - 8 | 1146 | 64 | ||
| GenBank mRNA | BC078388.1 | - | 6 - 7 | 686 | 64 | |
| EH459602.1 | - | 6 - 7 | 568 | 64 | ||
More Information
Set Membership: |
|

nadk2
Genomic Map