Assay Details
| Gene Symbol: caprin1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003484.1 | NP_001003484.1 | 11 - 12 | 1293 | 70 | |
| XM_005172274.4 | XP_005172331.1 | 11 - 12 | 1310 | 70 | ||
| XM_005172275.4 | XP_005172332.1 | 11 - 12 | 1310 | 70 | ||
| GenBank mRNA | BC078410.1 | - | 11 - 12 | 1293 | 70 | |
More Information
Set Membership: |
|

caprin1a
Genomic Map