Assay Details
| Gene Symbol: caprin1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003484.1 | NP_001003484.1 | 13 - 14 | 1440 | 87 | |
| XM_005172274.4 | XP_005172331.1 | 13 - 14 | 1457 | 87 | ||
| GenBank mRNA | BC078410.1 | - | 13 - 14 | 1440 | 87 | |
More Information
Set Membership: |
|

caprin1a
Genomic Map