Assay Details
| Gene Symbol: paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003504.1 | NP_001003504.1 | 5 - 6 | 813 | 62 | |
| GenBank mRNA | BC078339.1 | - | 5 - 6 | 813 | 62 | |
More Information
Set Membership: |
|

paip1
Genomic Map