Assay Details
| Gene Symbol: mipa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003534.1 | NP_001003534.1 | 3 - 4 | 991 | 70 | |
| GenBank mRNA | BC077129.1 | - | 3 - 4 | 991 | 70 | |
More Information
Set Membership: |
|

mipa
Genomic Map