Assay Details
| Gene Symbol: tagapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004548.2 | NP_001004548.2 | 3 - 4 | 252 | 81 | |
| NM_001365334.1 | NP_001352263.1 | 7 - 8 | 856 | 81 | ||
| GenBank mRNA | BC081595.1 | - | 3 - 4 | 252 | 81 | |
| EH448348.1 | - | 3 - 4 | 322 | 81 | ||
More Information
Set Membership: |
|

tagapb
Genomic Map