Assay Details
| Gene Symbol: tagapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004548.2 | NP_001004548.2 | 4 - 5 | 313 | 99 | |
| NM_001365334.1 | NP_001352263.1 | 8 - 9 | 917 | 99 | ||
| GenBank mRNA | BC081595.1 | - | 4 - 5 | 313 | 99 | |
| EH448348.1 | - | 4 - 5 | 383 | 99 | ||
More Information
Set Membership: |
|

tagapb
Genomic Map