Assay Details
| Gene Symbol: tagapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004548.2 | NP_001004548.2 | 7 - 8 | 749 | 71 | |
| NM_001365334.1 | NP_001352263.1 | 11 - 12 | 1353 | 71 | ||
| GenBank mRNA | BC081595.1 | - | 7 - 8 | 749 | 71 | |
More Information
Set Membership: |
|

tagapb
Genomic Map