Assay Details
| Gene Symbol: uck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004666.1 | NP_001004666.1 | 4 - 5 | 604 | 61 | |
| GenBank mRNA | BC081503.1 | - | 4 - 5 | 604 | 61 | |
More Information
Set Membership: |
|

uck1
Genomic Map