Assay Details
| Gene Symbol: trip10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007403.1 | NP_001007404.1 | 5 - 6 | 669 | 71 | |
| XM_017354074.2 | XP_017209563.1 | 5 - 6 | 702 | 71 | ||
| XM_017354077.2 | XP_017209566.1 | 5 - 6 | 700 | 71 | ||
| GenBank mRNA | BC085435.1 | - | 5 - 6 | 669 | 71 | |
| BC165714.1 | - | 5 - 6 | 438 | 71 | ||
More Information
Set Membership: |
|

trip10a
Genomic Map