Assay Details
| Gene Symbol: trip10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007403.1 | NP_001007404.1 | 8 - 9 | 1050 | 83 | |
| XM_017354074.2 | XP_017209563.1 | 8 - 9 | 1083 | 83 | ||
| XM_017354077.2 | XP_017209566.1 | 8 - 9 | 1081 | 83 | ||
| GenBank mRNA | BC085435.1 | - | 8 - 9 | 1050 | 83 | |
| BC165714.1 | - | 8 - 9 | 819 | 83 | ||
More Information
Set Membership: |
|

trip10a
Genomic Map