Assay Details
| Gene Symbol: mvda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007422.1 | NP_001007423.1 | 8 - 9 | 1120 | 150 | |
| GenBank mRNA | BC085325.1 | - | 8 - 9 | 1120 | 150 | |
More Information
Set Membership: |
|

mvda
Genomic Map