Assay Details
| Gene Symbol: pdzk1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007760.2 | NP_001007761.1 | 1 - 2 | 118 | 66 | |
| GenBank mRNA | BC133073.1 | - | 1 - 2 | 118 | 66 | |
More Information
Set Membership: |
|

pdzk1ip1
Genomic Map