Assay Details
| Gene Symbol: prkar1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009989.3 | NP_001009989.1 | 5 - 6 | 595 | 119 | |
| XM_005164023.3 | XP_005164080.1 | 5 - 6 | 629 | 119 | ||
| GenBank mRNA | BC088390.1 | - | 5 - 6 | 535 | 119 | |
| BC128799.1 | - | 5 - 6 | 578 | 119 | ||
More Information
Set Membership: |
|

prkar1aa
Genomic Map