Assay Details
| Gene Symbol: prkar1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009989.3 | NP_001009989.1 | 8 - 9 | 866 | 124 | |
| XM_005164023.3 | XP_005164080.1 | 8 - 9 | 900 | 124 | ||
| GenBank mRNA | BC088390.1 | - | 8 - 9 | 806 | 124 | |
| BC128799.1 | - | 8 - 9 | 849 | 124 | ||
More Information
Set Membership: |
|

prkar1aa
Genomic Map