Assay Details
| Gene Symbol: yes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013270.3 | NP_001013288.2 | 6 - 7 | 1102 | 71 | |
| GenBank mRNA | AJ620749.1 | - | 6 - 7 | 1084 | 71 | |
| BC128657.1 | - | 6 - 7 | 1107 | 71 | ||
More Information
Set Membership: |
|

yes1
Genomic Map