Assay Details
| Gene Symbol: camkk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014339.2 | NP_001014361.1 | 10 - 11 | 863 | 98 | |
| XM_005171848.4 | XP_005171905.1 | 12 - 13 | 2062 | 98 | ||
| XM_005171850.4 | XP_005171907.1 | 12 - 13 | 2061 | 98 | ||
| GenBank mRNA | BC091900.1 | - | 10 - 11 | 866 | 98 | |
More Information
Set Membership: |
|

camkk1a
Genomic Map