Assay Details
| Gene Symbol: camkk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014339.2 | NP_001014361.1 | 4 - 5 | 324 | 135 | |
| XM_005171848.4 | XP_005171905.1 | 6 - 7 | 1523 | 135 | ||
| XM_005171850.4 | XP_005171907.1 | 6 - 7 | 1522 | 135 | ||
| GenBank mRNA | BC091900.1 | - | 4 - 5 | 327 | 135 | |
More Information
Set Membership: |
|

camkk1a
Genomic Map