Assay Details
| Gene Symbol: camkk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014339.2 | NP_001014361.1 | 8 - 9 | 605 | 78 | |
| XM_005171848.4 | XP_005171905.1 | 10 - 11 | 1804 | 78 | ||
| XM_005171850.4 | XP_005171907.1 | 10 - 11 | 1803 | 78 | ||
| GenBank mRNA | BC091900.1 | - | 8 - 9 | 608 | 78 | |
More Information
Set Membership: |
|

camkk1a
Genomic Map