Assay Details
| Gene Symbol: lmbrd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017576.2 | NP_001017576.2 | 5 - 6 | 752 | 71 | |
| NM_001327914.1 | NP_001314843.1 | 5 - 6 | 752 | 71 | ||
| XM_009301180.3 | XP_009299455.1 | 5 - 6 | 750 | 71 | ||
| XM_021476149.1 | XP_021331824.1 | 5 - 6 | 1030 | 71 | ||
| XM_021476150.1 | XP_021331825.1 | 5 - 6 | 750 | 71 | ||
| GenBank mRNA | BC092730.1 | - | 5 - 6 | 710 | 71 | |
More Information
Set Membership: |
|

lmbrd2a
Genomic Map