Assay Details
| Gene Symbol: grap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017756.2 | NP_001017756.1 | 7 - 8 | 807 | 80 | |
| GenBank mRNA | BC093196.1 | - | 7 - 8 | 808 | 80 | |
| BC165551.1 | - | 5 - 6 | 608 | 80 | ||
More Information
Set Membership: |
|

grap2a
Genomic Map