Assay Details
| Gene Symbol: adss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017804.1 | NP_001017804.1 | 12 - 13 | 1326 | 96 | |
| GenBank mRNA | BC092877.1 | - | 12 - 13 | 1326 | 96 | |
More Information
Set Membership: |
|

adss2
Genomic Map