Assay Details
| Gene Symbol: stk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017839.1 | NP_001017839.1 | 7 - 8 | 1249 | 74 | |
| GenBank mRNA | BC092776.1 | - | 7 - 8 | 1249 | 74 | |
| BC165575.1 | - | 6 - 7 | 889 | 74 | ||
More Information
Set Membership: |
|

stk11
Genomic Map