Assay Details
| Gene Symbol: nmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020480.1 | NP_001018316.1 | 11 - 12 | 1456 | 82 | |
| GenBank mRNA | AB194986.1 | - | 11 - 12 | 1471 | 82 | |
| BC056805.1 | - | 12 - 13 | 1366 | 82 | ||
| BC095383.1 | - | 11 - 12 | 1456 | 82 | ||
More Information
Set Membership: |
|

nmt1a
Genomic Map