Assay Details
| Gene Symbol: rpp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020692.2 | NP_001018528.1 | 4 - 5 | 363 | 98 | |
| NR_144413.1 | 4 - 5 | 376 | 98 | |||
| XM_005173956.4 | XP_005174013.1 | 4 - 5 | 348 | 98 | ||
| GenBank mRNA | BC095792.1 | - | 4 - 5 | 333 | 98 | |
| BC165532.1 | - | 3 - 4 | 270 | 98 | ||
| EH542438.1 | - | 4 - 5 | 384 | 98 | ||
| EH595656.1 | - | 4 - 5 | 397 | 98 | ||
More Information
Set Membership: |
|

rpp14
Genomic Map