Assay Details
| Gene Symbol: arhgef3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020727.1 | NP_001018563.1 | 11 - 12 | 1422 | 116 | |
| XM_005162073.4 | XP_005162130.1 | 12 - 13 | 2101 | 116 | ||
| XM_005162074.4 | XP_005162131.1 | 13 - 14 | 2029 | 116 | ||
| GenBank mRNA | BC095043.1 | - | 11 - 12 | 1422 | 116 | |
| BC165626.1 | - | 11 - 12 | 1313 | 116 | ||
More Information
Set Membership: |
|

arhgef3l
Genomic Map