Assay Details
| Gene Symbol: arhgef3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020727.1 | NP_001018563.1 | 2 - 3 | 241 | 62 | |
| GenBank mRNA | BC095043.1 | - | 2 - 3 | 241 | 62 | |
| BC165626.1 | - | 2 - 3 | 132 | 62 | ||
More Information
Set Membership: |
|

arhgef3l
Genomic Map