Assay Details
| Gene Symbol: arhgef3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020727.1 | NP_001018563.1 | 3 - 4 | 288 | 66 | |
| XM_005162073.4 | XP_005162130.1 | 4 - 5 | 967 | 66 | ||
| XM_005162074.4 | XP_005162131.1 | 5 - 6 | 895 | 66 | ||
| GenBank mRNA | BC095043.1 | - | 3 - 4 | 288 | 66 | |
| BC165626.1 | - | 3 - 4 | 179 | 66 | ||
More Information
Set Membership: |
|

arhgef3l
Genomic Map