Assay Details
Gene Symbol: arhgef3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020727.1 | NP_001018563.1 | 9 - 10 | 1056 | 110 | |
XM_005162073.4 | XP_005162130.1 | 10 - 11 | 1735 | 110 | ||
XM_005162074.4 | XP_005162131.1 | 11 - 12 | 1663 | 110 | ||
GenBank mRNA | BC095043.1 | - | 9 - 10 | 1056 | 110 | |
BC165626.1 | - | 9 - 10 | 947 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
