Assay Details
| Gene Symbol: nelfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200168.1 | NP_956462.1 | 3 - 4 | 531 | 79 | |
| GenBank mRNA | AY391416.1 | - | 3 - 4 | 482 | 79 | |
| BC045409.1 | - | 3 - 4 | 531 | 79 | ||
| BC164663.1 | - | 3 - 4 | 396 | 79 | ||
More Information
Set Membership: |
|

nelfb
Genomic Map