Assay Details
| Gene Symbol: cnn3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024073.1 | NP_001019244.1 | 1 - 2 | 307 | 73 | |
| GenBank mRNA | BC107981.1 | - | 1 - 2 | 307 | 73 | |
More Information
Set Membership: |
|

cnn3b
Genomic Map